Tellur ≥99.999%, pulver -200 mesh
Leverantör: Thermo Scientific
Fara
11075.09 11075.18 11075.30
11075.09EA 2000 SEK
11075.09 11075.18 11075.30
Tellur ≥99.999%, pulver -200 mesh
Tellurium
Purity: 99.999% CAS No.: 13494-80-9 Physical Form: Powder Appearance: Grey Odour: Odourless UN No.: 3288 MDL No.: MFCD00134062
Formel: Te
MW: 127,6 g/mol
Kokpunkt: 990 °C (1013 hPa)
Smältpunkt: 450 °C
Densitet: 8,83 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134062
CAS nummer: 13494-80-9
EINECS: 236-813-4
UN: 3288
ADR: 6.1,III
Merck Index: 12,09273