Antimon ≥99.999%, spherical shots 1-5 mm (0.04-0.20 in) low oxide
Leverantör: Thermo Fisher Scientific
Varning
Ampuled under argon
38618.09 38618.18
38618.09EA 4100 SEK
38618.09 38618.18
Antimon ≥99.999%, spherical shots 1-5 mm (0.04-0.20 in) low oxide
Antimony
Purity: 99.999% CAS No.: 7440-36-0 Physical Form: Ampouled under argon Odour: Odourless MDL No.: MFCD00134030
Formel: Sb
MW: 121.76 g/mol
Kokpunkt: 1637
Smältpunkt: 630
Densitet: 6.684 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134030
CAS nummer: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1 III
Merck Index: 12,00733