Bly ≥99.998%, folie, Puratronic®, Thickness: 1.0 mm (0.04 in)
Leverantör: Thermo Scientific
Fara
12048.FF 12048.FI
12048.FFEA 1350 SEK
12048.FF 12048.FI
Bly ≥99.998%, folie, Puratronic®, Thickness: 1.0 mm (0.04 in)
Bly
Purity: 99.998% CAS No.: 7439-92-1 Physical Form: ≈7.1g/25x25mm Appearance: Grey Odour: Odourless MDL No.: MFCD00134050
Formel: Pb
MW: 207,2 g/mol
Kokpunkt: 1740 °C (1013 hPa)
Smältpunkt: 327 °C
Densitet: 11,3437 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134050
CAS nummer: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410