Bly 99,999 Metaller basis, rods, Ø 10 mm (0.39 in)
Leverantör: Thermo Fisher Scientific
Fara
11573.18 11573.30
11573.18EA 2500 SEK
11573.18 11573.30
Bly 99,999 Metaller basis, rods, Ø 10 mm (0.39 in)
Bly
Purity: 99.999% CAS No.: 7439-92-1 Physical Form: ≈5.6cm/50g, may be in multiple pieces Appearance: Grey Odour: Odourless MDL No.: MFCD00134050
Formel: Pb
MW: 207.2 g/mol
Kokpunkt: 1740
Smältpunkt: 327
Densitet: 11.3437 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134050
CAS nummer: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410