Tallium ≥99.99%, granulat ≤ 6 mm (0.2 in)
Leverantör: Thermo Scientific
Fara
Packaged in water.
12131.14 12131.22 12131.36
12131.14EA 6550 SEK
12131.14 12131.22 12131.36
Tallium ≥99.99%, granulat ≤ 6 mm (0.2 in)
Tallium
Purity: 99.99% CAS No.: 7440-28-0 Physical Form: Packaged in water Appearance: Blue-white Odour: Odourless UN No.: 1707 MDL No.: MFCD00134063
Formel: Tl
MW: 204,38 g/mol
Kokpunkt: 1457 °C (1013 hPa)
Smältpunkt: 301 °C
Densitet: 11,85 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134063
CAS nummer: 7440-28-0
EINECS: 231-138-1
UN: 1707
ADR: 6.1,II
Merck Index: 12,09391