Tallium 99,99 Metaller basis, rods, Ø 12.7 mm (0.5 in)
Leverantör: Thermo Fisher Scientific
Fara
00888.0Y 00888.DO
00888.0YEA 20225 SEK
00888.0Y 00888.DO
Tallium 99,99 Metaller basis, rods, Ø 12.7 mm (0.5 in)
Tallium
Purity: 99.99% CAS No.: 7440-28-0 Physical Form: ≈225g/150mm Odour: Odourless MDL No.: MFCD00134063
Formel: Tl
MW: 204.38 g/mol
Kokpunkt: 1457
Smältpunkt: 301
Densitet: 11.85 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134063
CAS nummer: 7440-28-0
EINECS: 231-138-1
UN: 1707
ADR: 6.1 II
Merck Index: 12,09391