Bor ≥99.4%, crystalline pieces 1-30 mm (0.039-1.2 in)
Leverantör: Thermo Scientific
Varning
41858.09 41858.18
41858.09EA 6225 SEK
41858.09 41858.18
Bor ≥99.4%, crystalline pieces 1-30 mm (0.039-1.2 in)
Boron
Purity: 99.4% CAS No.: 7440-42-8 Odour: Odourless MDL No.: MFCD00134034
Formel: B
MW: 10,81 g/mol
Densitet: 1,73 g/cm³ (15 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134034
CAS nummer: 7440-42-8
EINECS: 231-151-2
Merck Index: 13,01333