Strontium ≥99.95%, pieces, distilled dendritic
Leverantör: Thermo Fisher Scientific
Fara
Ampuled under argon
38624.06 38624.14
38624.06EA 20325 SEK
38624.06 38624.14
Strontium ≥99.95%, pieces, distilled dendritic
Strontium
Purity: 99.95% CAS No.: 7440-24-6 Physical Form: Ampouled under argon Sensitivity: Air and Moisture Sensitive UN No.: 3208 MDL No.: MFCD00134060
Formel: Sr
Kokpunkt: 1151 °C (1013 hPa)
Smältpunkt: 570…660 °C
Densitet: 2,38 g/cm³ (770 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134060
CAS nummer: 7440-24-6
EINECS: 231-133-4
UN: 3208
ADR: 4.3,II
Merck Index: 12,08994