Kalium 98%, chunks in mineral oil
Leverantör: Thermo Scientific
Fara
223860100 223861000 223860500 223865000
ACRO223860100EA 830 SEK
ACRO223860100 ACRO223861000 ACRO223860500 ACRO223865000
Kalium 98%, chunks in mineral oil
Kalium
Formel: K
MW: 39,1 g/mol
Kokpunkt: 770 °C (760 mmHg)
Smältpunkt: 63 °C
Densitet: 0,86 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00133776
CAS nummer: 7440-09-7
EINECS: 231-119-8
UN: 2257
ADR: 4.3,I
Merck Index: 12,07763
Order Now
Specifikation testresultat
Appearance Grey Chunks
Total trace metal impurities ≤2 %