2,2',4,4'-Tetrahydroxybenzophenone ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Bis(2,4-dihydroxyphenyl)methanone
A10237.14 A10237.22
A10237.14EA 1075 SEK
A10237.14 A10237.22
2,2',4,4'-Tetrahydroxybenzophenone ≥98%
2,2',4,4'-Tetrahydroxybenzophenone
Purity: 98+% CAS No.: 131-55-5 Molecular Formula: C13H10O5 Formula Weight: 246.22 Melting Point: 198-203° Beilstein Reference: 1914746
Formel: C₁₃H₁₀O₅
MW: 246,22 g/mol
Kokpunkt: 100…102 °C (40 mmHg)
Smältpunkt: 198…203 °C
Densitet: 1,216
Storage Temperature: Omgivande
MDL Number: MFCD00002278
CAS nummer: 131-55-5
EINECS: 205-028-9