1,3-Dichloro-4-(2-iodophenoxy)benzene

Leverantör: Thermo Scientific

CD08469DA CD08469EA CD08469EE
MAYBCD08469DAEA 0 SEK
MAYBCD08469DA MAYBCD08469EA MAYBCD08469EE
1,3-Dichloro-4-(2-iodophenoxy)benzene
1,3-Dichloro-4-(2-iodophenoxy)benzene
Formel: C₁₂H₇Cl₂IO
MW: 365 g/mol
CAS nummer: 175136-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR