(1S)-(-)-β-Pinene 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: (-)-Pin-2(10)-ene
150320010 150322500
ACRO150320010EA 1200 SEK
ACRO150320010 ACRO150322500
(1S)-(-)-β-Pinene 98%
(1S)-(-)-β-Pinene
Formel: C₁₀H₁₆
MW: 136.24 g/mol
Kokpunkt: 165…167 °C (760 mmHg)
Smältpunkt: –61 °C
Densitet: 0.87 g/cm³
Flampunkt: 32 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001345
CAS nummer: 18172-67-3
EINECS: 242-060-2
UN: 2319
ADR: 3 III
Merck Index: 12,07600
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4810 (20°C, 589 nm)
GC ≥97.5 %
Specific gravity (25°C) 0.860 to 0.880
Specific optical rotation -17° to -22° (25°C, 589 nm) (neat)