(1S)-(-)-β-Pinene 94,0 GC_ANALYSMETOD
Leverantör: TCI
|
Varning
|
Synonyms:
(-)-Pin-2(10)-ene
P0441-25ML
P0441-500ML
TCIAP0441-25MLEA
215
SEK
TCIAP0441-25ML
TCIAP0441-500ML
(1S)-(-)-β-Pinene 94,0 GC_ANALYSMETOD
(1S)-(-)-β-Pinene
Formel:
C₁₀H₁₆ MW: 136.24 g/mol Kokpunkt: 165°C Smältpunkt: -61 Flampunkt: 43°C |
MDL Number:
MFCD00063635 CAS nummer: 18172-67-3 EINECS: 242-060-2 UN: 3295 ADR: 3 III Merck Index: 14,07446 |
|
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 94.0 % |
Specific rotation [a]20/D | -19.0 to -23.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...