ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form), Millipore®
Leverantör: Merck Millipore Calbiochem
Synonyms: NADPH tetrasodium salt, Sodium (2R,3R,4R,5R)-2-(6-amino-9H-purin-9-yl)-5-((((((((2R,3S,4R,5R)-5-(3-carbamoylpyridin-1(4H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)oxidophosphoryl)oxy)methyl)-4-hydroxytetrahydrofuran-3-yl phosphate, Nicotinamide adenine dinucleotide phosphate tetrasodium salt, (reduced form)
481973-1 481973-100 481973-25 481973-50 481973-500
481973-1EA 29425 SEK
481973-1 481973-100 481973-25 481973-50 481973-500
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form), Millipore®
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)
Formel: C₂₁H₂₆N₇Na₄O₁₇P₃
MW: 833.35 g/mol
MDL Number: MFCD00036263
CAS nummer: 2646-71-1
EINECS: 220-163-3