Methyl violet 2B, ren, certified
Leverantör: Thermo Scientific
Varning
Synonyms: Basic Violet 1, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride
415070250 415071000 415075000
ACRO415070250EA 525 SEK
ACRO415070250 ACRO415071000 ACRO415075000
Methyl violet 2B, ren, certified
Methyl violet 2B
Formel: C₂₄H₂₈ClN₃
MW: 393,96 g/mol
Smältpunkt: 137 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012568
CAS nummer: 8004-87-3
UN: 3077
ADR: 9,III
Order Now
Specifikation testresultat
Appearance Green Crystalline powder
Infrared spectrum Conforms
Dye content ≥75 %
UV Lambda max 582 to 588 nm
Absorption Ratio P-15/P+15 1.06 to 1.19
BSC Certification Passes