Litmus indicator for titration (pH 5-8), Fluka™
Leverantör: Honeywell Chemicals
61280-25G 61280-25KG
61280-25GEA 3450 SEK
61280-25G HONC61280-25KG
Litmus indicator for titration (pH 5-8), Fluka™
Litmus
Storage Temperature: Omgivande
MDL Number: MFCD00131528
CAS nummer: 1393-92-6
EINECS: 215-739-6
Order Now
Specifikation testresultat
Change range (pH 5-8) complying