May Grunwald's eosin-methylene blue lösning, J.T.Baker®
Leverantör: Avantor
Fara
3855.1000EA 0 SEK
3855.1000 3855.2500
May Grunwald's eosin-methylene blue lösning, J.T.Baker®
May Grunwald's eosin-methylene blue
Storage Temperature: Omgivande
MDL Number: MFCD00131580
UN: 1230
ADR: 3,II
IVDLogo
Order Now
Specifikation testresultat
Performance Test passes test