May Grunwald's eosin-methylene blue, ren stain
Leverantör: Thermo Fisher Scientific
|
Varning
|
194830250
194831000
ACRO194830250EA
930
SEK
ACRO194830250
ACRO194831000
May Grunwald's eosin-methylene blue, ren stain
May Grunwald's eosin-methylene blue
Specifikation testresultat
Appearance | Dark green to dark brown Crystalline powder |
Infrared spectrum | Conforms |
Ratio | 1.2 to 2.1 Blue/red ratio |
UV Lambda max | 515 to 530 nm (red) |