Dodecylbenzene sulphonic acid mixture of C10-C13 isomers
Leverantör: Thermo Scientific
Fara
325900010 325905000
ACRO325900010EA 3825 SEK
ACRO325900010 ACRO325905000
Dodecylbenzene sulphonic acid mixture of C10-C13 isomers
Dodecylbenzene sulphonic acid mixture of C10-C13 isomers
Formel: C₁₈H₃₀O₃S
MW: 326,49 g/mol
Kokpunkt: 315 °C (1013 hPa)
Smältpunkt: –10 °C
Densitet: 1,06 g/cm³
Flampunkt: 210 °C
MDL Number: MFCD00066677
CAS nummer: 85536-14-7
EINECS: 248-289-4
UN: 2586
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Brown Viscous liquid
Infrared spectrum Conforms
Assay ≥96 % (Sulfonic acid, M.W. 322)
Water ≤0.5 % (K.F.)
Impurity ≤3 % Neutral oil
Sulfuric acid ≤1 %