Folin-Ciocalteu's reagent for analysis of phenols, Supelco®
Leverantör: Merck
Fara
Synonyms: Folin & Ciocalteu’s phenol reagent
1.09001.0100 1.09001.0500
1.09001.0100EA 735 SEK
1.09001.0100 1.09001.0500
Folin-Ciocalteu's reagent for analysis of phenols, Supelco®
Folin-Ciocalteu's reagent
Densitet: 1,22 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00132625
UN: 3264
ADR: 8,III
Order Now
Specifikation testresultat
Equivalent acid c(H+)= 2 mol/l (2N)
Sensitivity to phenol Conforms
Sensitivity to bovine serum albumin Conforms