Giemsa's stain lösning biological stain
Leverantör: Thermo Scientific
Fara
Synonyms: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue
295591000 295595000
ACRO295591000EA 435 SEK
ACRO295591000 ACRO295595000
Giemsa's stain lösning biological stain
Giemsa's stain
Densitet: 0,99 g/cm³
Flampunkt: 15 °C
Storage Temperature: Omgivande
MDL Number: MFCD00081642
CAS nummer: 51811-82-6
EINECS: 257-438-2
UN: 1230
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Dark blue to dark purple Solution
Additional info Giemsa stain powder in MeOH, Glycerol mixture
UV Lambda max 646 to 651 nm (c=0.1, MeOH) Methylene blue