X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt) 98%
Leverantör: Thermo Scientific
Synonyms: X-Gluc
331191000 331190250
ACRO331191000EA 7025 SEK
ACRO331191000 ACRO331190250
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt) 98%
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt)
Formel: C₂₀H₂₆BrClN₂O₇
MW: 521,79 g/mol
Smältpunkt: 230 °C
Storage Temperature: Frys
MDL Number: MFCD00058543
CAS nummer: 114162-64-0
Order Now
Specifikation testresultat
Appearance Colorless to white to off-white Powder or crystals or needles
Infrared spectrum Conforms
Water ≤5 % (K.F.)
HPLC ≥97.5 %