2,7-Diaminofluorene dihydrochloride

Leverantör: Merck

Synonyms: Fluorene-2,7-diyldiamine dihydrochloride, 2,7-Fluorenediammonium dichloride, Fluorene-2,7-diamine dihydrochloride

M-Clarity™ quality level = MQ200

8.20575.0001
8.20575.0001EA 1000 SEK
8.20575.0001
2,7-Diaminofluorene dihydrochloride
2,7-Diaminofluorene dihydrochloride
Formel: C₁₃H₁₄Cl₂N₂
MW: 269.17 g/mol
Smältpunkt: 260…310 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001128
CAS nummer: 13548-69-1
EINECS: 236-924-8
UN: 2811
Merck Index: 12,04191

Order Now

Specifikation testresultat

Solubility (20 °C) slightly soluble
Melting Point 260 - 310 °C
Molar Mass 269.17 g/mol
Synonyms 2,7-Diaminofluorene dihydrochloride
Hill Formula C13H14Cl2N2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR