(+)-Cinchonine, Sigma-Aldrich®
Leverantör: Merck
Varning
M-Clarity™ quality level = MQ200
8.02506.0025 8.02506.0100
8.02506.0025EA 945 SEK
8.02506.0025 8.02506.0100
(+)-Cinchonine, Sigma-Aldrich®
(+)-Cinchonine
Formel: C₁₉H₂₂N₂O
Smältpunkt: 261…264 °C
Densitet: 1,2 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00064372
CAS nummer: 118-10-5
EINECS: 204-234-6
Merck Index: 13,02308
Order Now
Specifikation testresultat
Solubility 0.25 g/l (20 °C)
Melting Point 258 - 264 °C
Molar Mass 294.40 g/mol
Bulk Density 130 kg/m3
pH 9.0 (0.25 g/l, H2O, 20 °C) (as aqueous solution)