L(-)-Menthol, EMPROVE® ESSENTIAL BP, Ph. Eur., JP, USP suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Varning
Synonyms: (1S,2R,4R)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, L-Menthol, Levomenthol, L(-)-Menthyl alcoholol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, (1R)-(-)-Menthol, (-)-Menthol, (1α,2β,4β)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, (1R,2S,5R)-(-)-Menthol
1.05995.1000 1.05995.9025
1.05995.1000EA 7025 SEK
1.05995.1000 1.05995.9025
L(-)-Menthol, EMPROVE® ESSENTIAL BP, Ph. Eur., JP, USP suitable for use as excipient, SAFC®
L(-)-Menthol
Formel: C₁₀H₂₀O
MW: 156.27 g/mol
Kokpunkt: 212
Smältpunkt: 41…44 °C
Densitet: 0.89 g/cm³ (20 °C)
Flampunkt: 96
Storage Temperature: Omgivande
MDL Number: MFCD00062979
CAS nummer: 2216-51-5
EINECS: 218-690-9
Merck Index: 13,05861