L(-)-Menthol 99.5%
Leverantör: Thermo Scientific
Varning
Synonyms: (1S,2R,4R)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, L-Menthol, Levomenthol, L(-)-Menthyl alcoholol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, (1R)-(-)-Menthol, (-)-Menthol, (1α,2β,4β)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, (1R,2S,5R)-(-)-Menthol
125401000 125405000 125400050 125400025
ACRO125401000EA 980 SEK
ACRO125401000 ACRO125405000 ACRO125400050 ACRO125400025
L(-)-Menthol 99.5%
L(-)-Menthol
Formel: C₁₀H₂₀O
MW: 156,27 g/mol
Kokpunkt: 212 °C (760 mmHg)
Smältpunkt: 41…43 °C
Densitet: 0,89 g/cm³
Flampunkt: 94 °C
Storage Temperature: Omgivande
MDL Number: MFCD00062979
CAS nummer: 2216-51-5
EINECS: 218-690-9
Merck Index: 13,05861
Order Now
Specifikation testresultat
Appearance Colorless to white Crystals or crystalline needles
Infrared spectrum Conforms
GC ≥99.4 %
Specific optical rotation -48° to -51° (20°C, 589 nm) (c=10, EtOH)