Disk- och desinficeringsmaskin, typ G 7883
Leverantör: MIELE
62.7883.46 5575720 7565630 5774540 5774530 5764960 5799600 5799550 5866740 5866780 5798620 5919760 5764950
141-1330EA 0 SEK
141-1330 141-1001 141-0582 141-0448 141-0447 141-0289 141-0462 141-0461 141-0286 141-0475 141-0454 141-0482 141-0288
Disk- och desinficeringsmaskin, typ G 7883
Diskmaskin för glasvaror
The G 7883 cleaner/disinfector is suitable for reprocessing laboratory glassware for analytical experiments in eg. industry, environmental and research laboratories. This appliance has a large two-level wash cabinet and can be integrated into a continuous worktop as a built-under unit or installed as a freestanding unit. After an intensive main cleaning phase, the disinfection takes place at >90 °C and a holding time of 5 minutes. The final rinsing is carried out with deionised water without rinsing agents.
  • 10 standard cleaning programmes
  • Main wash/rinse cycle optionally with fully demineralised water
  • Thermal disinfection
  • Electrical door lock
  • With water softener inside and steam condenser
  • Programme safety cut-out
  • Optical and acoustic signal at end of programme
  • Hygienic freshwater system with fresh water intake for each programme stage
  • 3 spray arms for thorough cleaning of laboratory glassware
  • Thorough cleaning of lumens with injector system
  • Flowmeter to monitor water intake quantities
Greater capacity - Increased purity - More flexibility.
Certifikat: Uppfyller VDE, skyddad mot EMC/radiostörningar, DVGW, IP X1.
Beställningsinformation: Korgarna beställs separat. Andra modeller, t ex med vitt hölje eller med kantdekor, finns tillgängliga vid förfrågan. Vid användning som fristående enhet måste topplocket beställas separat.