HD Super™ process tables
Leverantör: Metro International
WT309US WT367HS MWT309US WT369US WT366US MWT307US
132-2106EA 32025 SEK
132-2106 METOWT367HS 132-0960 132-0962 METOWT366US METOMWT307US
HD Super™ process tables
Inredning Bord
These stainless steel laboratory workbenches are available in two versions, with or without a bottom shelf. The versions without a bottom shelf have three transverse struts allowing free footroom without an impeding front bar.
  • Unobstructed flat surface for unimpeded work
  • No sharp edges through specially rounded corners
  • All stationary tables are 864 mm high with stationary posts and levelling feet (feet individually height-adjustable)
  • All mobile tables are 864 mm high with special posts and 4 poly casters (127 mm) - 2 swibel/2 brake