Mikrocentrifug, med eller utan kyla, 5424 / 5424R (IVD)
Leverantör: EPPENDORF
5424000.061 5424000.266 5424000.460 5424000.665 5404000.065 5404000.260 5404000.464 5404000.669
521-0730EA 0 SEK
521-0730 521-0732 521-0734 521-0736 521-1540 521-1541 521-1542 521-1543
Mikrocentrifug, med eller utan kyla, 5424 / 5424R (IVD)
Centrifuger Mikrocentrifuger
Microcentrifuges with 24 positions, extremely quiet running and perfectly designed for use in all biology and microbiology labs. Available in two versions: with rotary control knobs or touch sensitive keypad.
  • Rotary knob version for fast parameter selection
  • Keypad version for easy cleaning
  • Soft-touch one-finger lid closure for ergonomic operation
  • OptiBowl® design for quiet operation, even without a rotor lid
  • Centrifuge 5424 R cools down to 4 °C in only 8 minutes
Beställningsinformation: Levereras med standardrotor FA-45-24-11 (24×1,5/2,0 ml) med lock.