Tillbehör för termoreaktorer Spectroquant® TR-serien
Leverantör: Merck
1.71203.0001 1.71204.0001
1.71203.0001EA 5560 SEK
1.71203.0001 1.71204.0001
Tillbehör för termoreaktorer Spectroquant® TR-serien
Blocktermostater Blocktermostater, digitala
The thermosensor measures the current temperature in the hole of the thermoreactor block and compares it with a specified temperature. The results can be transmitted to a PC for documentation purposes. Optimised, well matched system.