pH-elektrod för Knick® mätare
Leverantör: HAMILTON BONADUZ
242066 242068 242070
662-9230EA 4325 SEK
662-9230 662-9235 662-9243
pH-elektrod för Knick® mätare
Elektroder pH-elektroder
These electrodes are especially well suited for Knick® Portamess® equipment. The electrode head creates a hermetic seal with the Portamess® storage tube.
  • Liq-Glass Knick Temp DIN: Everef reference system provides stable potentials resulting in long sensor lifetime
  • Double Pore Knick/Polyplast Knick Temp DIN: Ag/AgCl reference system
  • Specially formulated electrolytes allow pH measurements of aqueous and biological (protein containing) matrices
  • Special diaphragm shapes for small sample volumes or semi-solid samples
pH-område: 0 till 14
* Inbyggd temperaturgivare
Beställningsinformation: Levereras med individuellt certifikat och serienummer.