VWR® PUREImage, Anteckningsbok, för renrum
Leverantör: VWR Collection
818-0126EA 2800 SEK
818-0126 818-0127 818-0128
VWR® PUREImage, Anteckningsbok, för renrum
Anteckningsböcker
These notebooks contain VWR® PUREImage, latex-free paper and have a see-through cover. Designed for use in cleanrooms and critical environments, where particulate or extractable contamination control is imperative.
  • Produced with new, latex-free, paper-binding technology
  • College ruled on all pages
  • Double-bagged in a Class 100 environment
Format: A4, A5, A6, 100 sidor (50 blad).