Mätpipett, klass AS
Leverantör: HIRSCHMANN
1101129 1101133 1101135 1101140 1101141 1101142 1101153 1101154 1101160 1101165 1101170 1101175
612-1033EA 120 SEK
612-1033 612-1034 612-1035 612-1036 612-1037 612-1038 612-1039 612-1040 612-1041 612-1042 612-1043 612-1044
Mätpipett, klass AS
Pipetter Pipetter, graderade
AR-Glas®.
  • Calibrated to deliver 'Ex', certificate of conformity with dated batch identification available on internet (www.hirschmannlab.de)
  • Marks and inscriptions in blue, graduated to the tip
  • From 5 ml with a cotton plug as standard
  • Waiting time five seconds
DIN EN ISO 835
Length: 360 mm ±5 mm except for 25 and 50 ml pipettes: 450 mm ±5 mm.
Certifikat: DIN EN ISO 835. Conformity certified (DE-M marking).