Mätpipett, klass AS
Leverantör: HIRSCHMANN
1100229 1100233 1100240 1100241 1100242 1100253 1100260 1100270
612-2766EA 125 SEK
612-2766 612-2767 612-2768 612-2769 612-2770 612-2771 612-2772 612-2773
Mätpipett, klass AS
Pipetter Pipetter, graderade
Glass.
  • Calibrated to deliver 'Ex', conformity certified with dated batch identification
  • Marks and inscriptions in amber stain, not graduated to tip (partial delivery)
  • From 5 ml with a cotton plug upper end
DIN 12 697
Längd: 360 ±5 mm utom pipetter på 25 ml: 450 ±5 mm
Certifikat: DIN EN ISO 835. Conformity certified (DE-M marking).