Mätpipett, typ 3, klass AS, justerad för full tömning
Leverantör: HIRSCHMANN
1100129 1100133 1100135 1100140 1100141 1100142 1100153 1100154 1100160 1100165 1100170 1100175 110015308 110015345 110013327 110014127 110015327 110016027 110017027 1100165DKD 1100175DKD 1100154DKD 1100160DKD 1100135DKD 1100142DKD 1100170DKD
612-1045EA 125 SEK
612-1045 612-1046 613-2111 613-2112 612-1047 613-2113 612-1048 612-1049 612-1050 612-1051 612-1052 612-1053 612-7283 612-5751 612-4723 612-4727 612-4731 612-4735 612-4744 612-4743 612-4746 612-4733 612-4742 612-4725 612-4729 612-4745
Mätpipett, typ 3, klass AS, justerad för full tömning
Pipetter Pipetter, graderade
AR-Glas®.
  • Calibrated to deliver 'Ex', certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
  • Marks and inscriptions in amber stain, graduated to the tip
  • From 5 ml with a cotton plug as standard
  • Waiting time five seconds
DIN EN ISO 835
Length: 360 mm ±5 mm except for 25 and 50 ml pipettes: 450 mm ±5 mm
Certifikat: DIN EN ISO 835. Conformity certified (DE-M marking).