Mätcylindrar, hög form, klass B, borosilikatglas, SILBERBRAND ETERNA
Leverantör: Brand
31962 31964 31920 31905 31908 31928 31938 31948 31954
612-5014EA 930 SEK
612-5014 612-4218 612-5003 612-5000 612-5002 612-5004 612-5011 612-5012 612-5013
Mätcylindrar, hög form, klass B, borosilikatglas, SILBERBRAND ETERNA
Cylindrar Measuring Cylinders
Borosilikatglas 3.3, hög modell, klass B.
  • Calibrated to contain (TC, In)
  • Hexagonal base, spout
  • Short graduation marks
  • Marks and inscriptions in ETERNA amber stain
DIN EN ISO 4788
Certifikat: DIN EN ISO 4788.