Blandande mätcylindrar, hög form, klass A, borosilikatglas, BLAUBRAND®
Leverantör: Brand
32462 32438 32448 32408 32420 32428 32454 932408
612-5029EA 3175 SEK
612-5029 612-5026 612-5027 612-5023 612-5024 612-5025 612-5028 BRND932408
Blandande mätcylindrar, hög form, klass A, borosilikatglas, BLAUBRAND®
Cylindrar Measuring Cylinders
Borosilicate glass 3.3, class A, conformity certified.
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, PP stopper (except for 612-5028 and 612-5029 octagonal PE stopper)
  • Ring marks at major graduations, marks and inscriptions in high contrast blue enamel
  • Total order from one batch
* Höjd utan propp
Certifikat: DIN EN ISO 4788.
Beställningsinformation: Supplied with a batch certificate. Measuring cylinders are also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.