Separertratt, Squibb, Nalgene®
Leverantör: Thermo Fisher Scientific
4300-0125 4300-0250 4300-0500 4300-1000
527-0005EA 1875 SEK
527-0005 527-0006 527-0007 527-0008
Separertratt, Squibb, Nalgene®
Trattar Separertrattar
PPCO translucent, TFE stopcock, PP screw closure.
  • Leakproof closure and stopcock
  • Very good chemical resistance
  • Autoclavable except for stopcock assembly
  • TFE stopcock requires no lubrication