Filtrertratt enligt Allihn, VitraPOR®
Leverantör: ROBU GLASFILTER
30301 30302 30303 30304 30300
511-3101EA 590 SEK
511-3101 511-3102 511-3103 511-3104 511-0241
Filtrertratt enligt Allihn, VitraPOR®
Trattar Filtertrattar Mikrofiltertrattar
Borsilikatglas 3,3.
  • According to Allihn
  • Micro-scale filtration
  • Integrated VitraPOR® filter
  • Minimal dead volume
  • High chemical resistance and reusable
Med integrerad glasfilterplatta. Höjd över glasfilterplatta: 100 mm.
The Allihn-style design ensures minimal dead volume and optimal flow dynamics. Made of borosilicate glass, the funnel is highly resistant to thermal and chemical stress, making it ideal for routine use in demanding environments.
Certifikat:
In compliance with ISO 4796-80 and ASTM E128-99 standards.