Flasks, round bottom with Quickfit® ground joints
Leverantör: SciLabware
FR500/3U FR50/3M FR100/3M FR250/3M FR500/3M FR150/3M FR1L/3UM
212-0613EA 320 SEK
212-0613 201-8532 212-0605 201-8502 201-8535 BIBBFR150/3M 214-0418
Flasks, round bottom with Quickfit® ground joints
Kolvar Kolv, rund
PYREX®, borosilicate glass, medium neck.
  • With a longer neck than standard Quickfit® flasks
  • Length of neck: 40 mm (1 L) and 45 mm (remaining)
Useful for reducing problems associated with frothing and splashing.
Certifikat: ISO 1773, BS 6352