Jars with screw cap
Leverantör: Anicrin
C150P121 C150P220 C150P221 C150P120 60.40.18BCAP C150P210 C150P211 C150P111
216-2050EA 1200 SEK
216-2050 216-2051 216-2052 216-2049 216-1570 216-2047 216-2048 216-2029
Jars with screw cap
Burkar Burkar, universal
Slightly conical vessels, made of PP, translucent with red PE screw cap, for sampling, storage and transportation.
  • Twist cap for quick opening and closing
  • Very good temperature resistance
  • Very good chemical resistance
Observera: *Produkten/produkterna på denna sida ligger utanför tillämpningsområdet för EU -direktivet 2019/904