Slangkoppling, L-formad
Leverantör: KARTELL
71250 71251 71252 71253 71254 71255 71256
229-0735EA 135 SEK
229-0735 229-0736 229-0737 229-0738 229-0739 229-0740 229-0741
Slangkoppling, L-formad
Beslag och kopplingar
PP, genomskinlig.
  • Good mechanical strength and chemical resistance
  • Very good thermal resistance
  • Autoclavable
  • Suitable for foodstuff
Dimensions are shown as both nominal and exact as elasticity of tubing will influence the choice of size. Use our tubing selector 229-0111 to determine 'grip'. This will then give equivalent crest dimension for the particular connector concerned.
They meet with the Food and Drug regulations.90/128CEE 92/39CEE and FDA 21 CFR 177.1520.
Suitable for foodstuff.
Certifikat: Conforms to Food and Drug regulations 90/128 CEE, 92/39 CEE and FDA 21 CFR 177.1520.