Slangkoppling, Y-formad
Leverantör: KARTELL
70527 70528 70529 70530 70531 70532
229-0723EA 265 SEK
229-0723 229-0724 229-0725 229-0726 229-0727 229-0728
Slangkoppling, Y-formad
Beslag och kopplingar
PP, vit.
  • Good mechanical strength and chemical resistance
  • Very good thermal resistance
  • Autoclavable
  • Suitable for foodstuff
Med avsmalnande ändar.
Certifikat: Conforms to Food and Drug regulations 90/128 CEE, 92/39 CEE and FDA 21 CFR 177.1520.