Screw caps, for DIN 168 bottles
Leverantör: KARTELL
15800 15840 15880 16120 16160
217-0483EA 650 SEK
217-0483 217-0484 217-0485 KART16120 KART16160
Screw caps, for DIN 168 bottles
Lock Lock, flaskor
HDPE.
Standard screw caps for polyethylene bottles manufactured to DIN 168.