Byrett, rak kran
Leverantör: HIRSCHMANN
3130160 3130170 3130175
612-3544EA 995 SEK
612-3544 612-3546 612-3547
Byrett, rak kran
Byretter
DURAN®, borosilicate glass, class AS.
  • Calibrated to deliver 'Ex', conformity certified (DE-M marking), with dated batch identification
  • Ring marks at major graduations, black graduations
  • With glass key
Skallängd: 500 mm.
Scale length: 500 mm
Certifikat: DIN ISO 385.