Byrett, rak kran, Schellbach
Leverantör: HIRSCHMANN
3140160 3140170 3140175 3140260 3140270 3140275 3140360 3140370 3140375
612-9811EA 1025 SEK
612-9811 612-9812 612-9813 612-3552 612-3553 612-3554 612-5757 612-5758 612-5759
Byrett, rak kran, Schellbach
Byretter
DURAN®, borosilicate glass, class AS.
  • Calibrated to deliver 'Ex', conformity certified (DE-M marking) with dated batch identification
  • Schellbach stripe, ring marks at major graduations, blue graduations
Skallängd: 500 mm
Scale length: 500 mm
Certifikat: DIN EN ISO 385