Carboys, smal hals, PC, med skruvlock, Nalgene®
Leverantör: Thermo Fisher Scientific
2251-0020 2251-0050
216-8781EA 2400 SEK
216-8781 216-8782
Carboys, smal hals, PC, med skruvlock, Nalgene®
Flaskor Dunkar
PC, transparent with white PP screw cap.
  • God kemikalieresistens
  • Tål temperaturer från ‒100 upp till +135 °C, autoklaverbar
  • Lätt och robust, graderad
Can be used in any situation. Particularly suitable for the production of large quantities of nutrient and culture media, particularly for cultures that require light, e.g. algae. Ideal for storing and dispensing reagents and distilled water. Can be used in cold rooms and at temperatures below freezing.
Conduct large-volume media and culture preparation with these polycarbonate Nalgene Clearboys, particularly when visual inspection is required. This polycarbonate Clearboy is durable and safer than glass, autoclavable for sterile applications and ideal for refrigerated or frozen storage of aqueous solutions such as sterile water, aqueous reagents and media.
Leveransinformation: Includes polypropylene closure and TPE gasket.