Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 M i vattenlösning
Leverantör: Thermo Fisher Scientific
Synonyms: Tris(1,10-phenanthroline)iron (II) sulphate, Tris(1,10-phenanthroline-N1,N10)iron sulphate
30562.18 30562.30
30562.18EA 1425 SEK
30562.18 30562.30
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 M i vattenlösning
Ferroin (1,10-phenanthrolineferrous sulphate complex)
CAS No.: 14634-91-4 Molecular Formula: C36H24FeN6O4S Formula Weight: 692.53 Physical Form: Liquid Appearance: Red
Formel: C₃₆H₂₄FeN₆SO₄
MW: 692.54 g/mol
Densitet: 1.01 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00036428
CAS nummer: 14634-91-4
EINECS: 238-676-6