Pararosaniline hydrochloride, pure, high purity biological stain
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Basic Red 9
227880250 227881000 227880050 227885000
ACRO227880250EA 1550 SEK
ACRO227880250 ACRO227881000 ACRO227880050 ACRO227885000
Pararosaniline hydrochloride, pure, high purity biological stain
Pararosaniline hydrochloride
Formel: C₁₉H₁₈ClN₃
MW: 323.82 g/mol
Smältpunkt: 268…270 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00001657
CAS nummer: 569-61-9
EINECS: 209-321-2
Order Now
Specifikation testresultat
Appearance Green Crystalline powder
Infrared spectrum Conforms
Dye content ≥85 %