1,2,4-Triazol-4-ylamin 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Amino-1,2,4-triazole, 1,2,4-Triazol-4-ylamine, 4H-1,2,4-Triazol-4-amine
104700250 104701000
ACRO104700250EA 1700 SEK
ACRO104700250 ACRO104701000
1,2,4-Triazol-4-ylamin 99%
1,2,4-Triazol-4-ylamin
Formel: C₂H₄N₄
MW: 84,08 g/mol
Smältpunkt: 84 °C
Flampunkt: >149 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003099
CAS nummer: 584-13-4
EINECS: 209-533-5
Order Now
Specifikation testresultat
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
HPLC ≥98.5 %
UV at 300 nm A: ≤0.005
Residue after evaporation ≤2 ppm